Kalium(3-(methansulfonylamino)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium trifluoro(3-(methylsulfonamido)phenyl)borate

TB-3236-1G TB-3236-5G
COBBTB-3236-1GEA 310 EUR
COBBTB-3236-1G COBBTB-3236-5G
Kalium(3-(methansulfonylamino)phenyl)trifluoroborat
Kalium(3-(methansulfonylamino)phenyl)trifluoroborat
Formel: C₇H₈BF₃KNO₂S
Molecular Weight: 277,11 g/mol
MDL: MFCD04115762
CAS-Nummer: 850623-67-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR