Kalium(3,5-dinitro-2-methylphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3242-1G TB-3242-25G TB-3242-5G
COBBTB-3242-1GEA 98.3 EUR
COBBTB-3242-1G COBBTB-3242-25G COBBTB-3242-5G
Kalium(3,5-dinitro-2-methylphenyl)trifluoroborat
Potassium-3,5-dinitro-2-methylphenyltrifluoroborate
Formel: C₇H₅BF₃KN₂O₄
Molecular Weight: 288.03 g/mol
MDL: MFCD04115767
CAS-Nummer: 850623-72-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR