Kalium(3-pyridyl)trifluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Potassium trifluoro(pyridin-3-yl)borate

711101-1G 711101-250MG
SIAL711101-1GEA 96.5 EUR
SIAL711101-1G SIAL711101-250MG
Kalium(3-pyridyl)trifluoroborat, Sigma-Aldrich®
Potassium(3-pyridyl)trifluoroborate
Formel: C₅H₄BF₃KN
Molecular Weight: 185,0000 g/mol
MDL: MFCD09038545
CAS-Nummer: 561328-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR