Kalium(2-fluor-6-methyl-3-pyridyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
Potassium trifluoro(2-fluoro-6-methylpyridin-3-yl)borate
TB-0012-1G
TB-0012-5G
COBBTB-0012-1GEA
303
EUR
COBBTB-0012-1G
COBBTB-0012-5G
Kalium(2-fluor-6-methyl-3-pyridyl)trifluoroborat
Potassium(2-fluoro-6-methyl-3-pyridyl)trifluoroborate
Formel:
C₆H₅BF₄KN Molecular Weight: 217,01 g/mol |
MDL:
MFCD09260453 CAS-Nummer: 1098173-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...