Bis(trimethylsilyl)sulfat
Lieferant: CHEMPUR SILANE
Synonyme:
Bis(trimethylsilyl) sulphate
SILS02810.5
SILS02810.25
SILS02810.100
SILNSILS02810.5EA
0
EUR
SILNSILS02810.5
SILNSILS02810.25
SILNSILS02810.100
Bis(trimethylsilyl)sulfat
Bis(trimethylsilyl)sulfat
Formel:
C₆H₁₈O₄SSi₂ Molecular Weight: 242.4407 g/mol |
MDL:
MFCD00008268 CAS-Nummer: 18306-29-1 EINECS: 242-192-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...