4-Formyltetrahydropyran
Lieferant: COMBI-BLOCKS
Synonyme:
Tetrahydropyran-4-carbaldehyde, Tetrahydro-2H-pyran-4-carboxaldehyde
AM-1016-1G
AM-1016-5G
COBBAM-1016-1GEA
103
EUR
COBBAM-1016-1G
COBBAM-1016-5G
4-Formyltetrahydropyran
4-Formyltetrahydropyran
Formel:
C₆H₁₀O₂ Molecular Weight: 114.1424 g/mol |
MDL:
MFCD03425264 CAS-Nummer: 50675-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...