2-(Methoxycarbonyl)-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate

PN-5085-1G PN-5085-250M
COBBPN-5085-1GEA 596 EUR
COBBPN-5085-1G COBBPN-5085-250M
2-(Methoxycarbonyl)-4-pyridinboronsäurepinakolester
2-(Methoxycarbonyl)-4-pyridinboronsäurepinakolester
Formel: C₁₃H₁₈BNO₄
Molecular Weight: 263.1000 g/mol
MDL: MFCD08752647
CAS-Nummer: 957062-72-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR