2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-5-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)pyridine

PN-9116-1G PN-9116-250M
COBBPN-9116-1GEA 334 EUR
COBBPN-9116-1G COBBPN-9116-250M
2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-5-boronsäurepinakolester
2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-5-boronsäurepinakolester
Formel: C₁₆H₂₄BNO₄
Molecular Weight: 305.18 g/mol
MDL: MFCD09702390
CAS-Nummer: 910036-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR