5-Brom-3-nitro-2-pyridincarbonitril

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-cyano-3-nitropyridine, 5-Bromo-3-nitropicolinonitrile, 5-Bromo-3-nitropyridine-2-carbonitrile

OS-7673-5G OS-7673-25G OS-7673-100G
COBBOS-7673-5GEA 37.5 EUR
COBBOS-7673-5G COBBOS-7673-25G COBBOS-7673-100G
5-Brom-3-nitro-2-pyridincarbonitril
5-Brom-3-nitro-2-pyridincarbonitril
Formel: C₆H₂BrN₃O₂
Molecular Weight: 228,0030 g/mol
MDL: MFCD06657551
CAS-Nummer: 573675-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR