5-Brom-3-nitro-2-pyridincarbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromo-2-cyano-3-nitropyridine, 5-Bromo-3-nitropicolinonitrile, 5-Bromo-3-nitropyridine-2-carbonitrile

M-Clarity™ Qualitätsstufe = MQ100

662968-25G 662968-5G
SIAL662968-25GEA 121 EUR
SIAL662968-25G SIAL662968-5G
5-Brom-3-nitro-2-pyridincarbonitril, Sigma-Aldrich®
5-Brom-3-nitro-2-pyridincarbonitril
Formel: C₆H₂BrN₃O₂
Molecular Weight: 228,0030 g/mol
MDL: MFCD06657551
CAS-Nummer: 573675-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR