6-Acetamido-3-brompicolinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
6-Acetamido-3-bromopyridine-2-carboxylic acid
CA-4146-5G
CA-4146-1G
COBBCA-4146-5GEA
0
EUR
COBBCA-4146-5G
COBBCA-4146-1G
6-Acetamido-3-brompicolinsäure
6-Acetamido-3-brompicolinsäure
Formel:
C₈H₇BrN₂O₃ |
MDL:
MFCD08235101 CAS-Nummer: 882430-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...