3-Brom-5-(methylthio)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-methylthiopyridine

OT-0103-1G OT-0103-5G
COBBOT-0103-1GEA 227 EUR
COBBOT-0103-1G COBBOT-0103-5G
3-Brom-5-(methylthio)pyridin
3-Bromo-5-(methylthio)pyridine
Formel: C₆H₆BrNS
MDL: MFCD08689537
CAS-Nummer: 142137-18-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR