5-Brom-2-(methylthio)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-methylthiopyridine, 2-Methylthio-5-bromopyridine

PY-7790-1G PY-7790-5G
COBBPY-7790-1GEA 75.8 EUR
COBBPY-7790-1G COBBPY-7790-5G
5-Brom-2-(methylthio)pyridin
5-Brom-2-(methylthio)pyridin
Formel: C₆H₆BrNS
Molecular Weight: 204.0875 g/mol
MDL: MFCD03411571
CAS-Nummer: 51933-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR