Methyl-3-(Boc-amino)picolinat

Lieferant: COMBI-BLOCKS

PY-1327-5G PY-1327-1G
COBBPY-1327-5GEA 884 EUR
COBBPY-1327-5G COBBPY-1327-1G
Methyl-3-(Boc-amino)picolinat
Methyl-3-(Boc-amino)picolinat
Formel: C₁₂H₁₆N₂O₄
MDL: MFCD12026422
CAS-Nummer: 912369-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR