3-(Piperidin-1-ylsulfonyl)pyridin

Lieferant: COMBI-BLOCKS

PY-7051-1G PY-7051-5G
COBBPY-7051-1GEA 303 EUR
COBBPY-7051-1G COBBPY-7051-5G
3-(Piperidin-1-ylsulfonyl)pyridin
3-(Piperidin-1-ylsulfonyl)pyridin
Formel: C₁₀H₁₄N₂O₂S
MDL: MFCD05149384
CAS-Nummer: 26103-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR