3-Acetyl-2-brompyridin

Lieferant: CHEMPUR

10-0019.1 10-0019.5
CHMP10-0019.1EA 0 EUR
CHMP10-0019.1 CHMP10-0019.5
3-Acetyl-2-brompyridin
3-Acetyl-2-brompyridin
Formel: C₇H₆BrNO
Molecular Weight: 200,0300 g/mol
MDL: MFCD04114370
CAS-Nummer: 84199-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR