5-Brom-2-hydroxy-4-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-4-(trifluoromethyl)pyridin-2-ol

PY-1153-5G PY-1153-1G
COBBPY-1153-5GEA 995 EUR
COBBPY-1153-5G COBBPY-1153-1G
5-Brom-2-hydroxy-4-(trifluormethyl)pyridin
5-Brom-2-hydroxy-4-(trifluormethyl)pyridin
Formel: C₆H₃BrF₃NO
Molecular Weight: 241,9933 g/mol
MDL: MFCD11227189
CAS-Nummer: 109919-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR