3-Brom-2-(methylthio)-5-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

PY-1420-1G PY-1420-5G
COBBPY-1420-1GEA 204 EUR
COBBPY-1420-1G COBBPY-1420-5G
3-Brom-2-(methylthio)-5-(trifluormethyl)pyridin
3-Bromo-2-(methylthio)-5-(trifluoromethyl)pyridine
Formel: C₇H₅BrF₃NS
Molecular Weight: 272,0855 g/mol
MDL: MFCD16295130
CAS-Nummer: 1226808-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR