5-Brom-4-methyl-2-(methylamino)pyridin

Lieferant: COMBI-BLOCKS

PY-1229-1G PY-1229-5G PY-1229-250M
COBBPY-1229-1GEA 484 EUR
COBBPY-1229-1G COBBPY-1229-5G COBBPY-1229-250M
5-Brom-4-methyl-2-(methylamino)pyridin
5-Brom-4-methyl-2-(methylamino)pyridin
Formel: C₇H₉BrN₂
MDL: MFCD07375098
CAS-Nummer: 155789-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR