5-Bromo-2-methylamino-3-nitro-4-picoline

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-4-methyl-2-methylamino-3-nitropyridine, 5-Bromo-N,4-dimethyl-3-nitropyridin-2-amine

PY-1294-1G PY-1294-5G
COBBPY-1294-1GEA 137 EUR
COBBPY-1294-1G COBBPY-1294-5G
5-Bromo-2-methylamino-3-nitro-4-picoline
5-Bromo-2-methylamino-3-nitro-4-picoline
Formel: C₇H₈BrN₃O₂
Molecular Weight: 246,1 g/mol
MDL: MFCD08692210
CAS-Nummer: 155790-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR