5-Brom-2-methoxy-4-methyl-3-nitropyridin

Lieferant: COMBI-BLOCKS

PY-1276-1G PY-1276-25G PY-1276-5G
COBBPY-1276-1GEA 65.8 EUR
COBBPY-1276-1G COBBPY-1276-25G COBBPY-1276-5G
5-Brom-2-methoxy-4-methyl-3-nitropyridin
5-Brom-2-methoxy-4-methyl-3-nitropyridin
Formel: C₇H₇BrN₂O₃
Molecular Weight: 247.0500 g/mol
MDL: MFCD06659504
CAS-Nummer: 884495-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR