2-(Ethylamino)-3-nitro-5-picolin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Ethylamino-5-methyl-3-nitropyridine

PY-7387-1G PY-7387-5G
COBBPY-7387-1GEA 49.7 EUR
COBBPY-7387-1G COBBPY-7387-5G
2-(Ethylamino)-3-nitro-5-picolin
2-(Ethylamino)-3-nitro-5-picolin
Formel: C₈H₁₁N₃O₂
MDL: MFCD09862688
CAS-Nummer: 106690-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR