2-(Cyclohexylamino)-3-nitro-5-picolin

Lieferant: COMBI-BLOCKS

PY-7385-1G PY-7385-5G
COBBPY-7385-1GEA 74.2 EUR
COBBPY-7385-1G COBBPY-7385-5G
2-(Cyclohexylamino)-3-nitro-5-picolin
2-(Cyclohexylamino)-3-nitro-5-picolin
Formel: C₁₂H₁₇N₃O₂
MDL: MFCD10699716
CAS-Nummer: 1033202-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR