2-(Cyclopropylamino)-3-nitro-5-picolin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Cyclopropylamino-5-methyl-3-nitropyridine

PY-7384-1G PY-7384-5G
COBBPY-7384-1GEA 124 EUR
COBBPY-7384-1G COBBPY-7384-5G
2-(Cyclopropylamino)-3-nitro-5-picolin
2-(Cyclopropylamino)-3-nitro-5-picolin
Formel: C₉H₁₁N₃O₂
MDL: MFCD10699715
CAS-Nummer: 1033202-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR