3-Methyl-4-nitropyridin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Nitro-3-picolin

OS-7550-5G OS-7550-1G
COBBOS-7550-5GEA 111 EUR
COBBOS-7550-5G COBBOS-7550-1G
3-Methyl-4-nitropyridin
3-Methyl-4-nitropyridin
Formel: C₆H₆N₂O₂
Molecular Weight: 138,12 g/mol
MDL: MFCD03095084
CAS-Nummer: 1678-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR