2,6-Bis(brommethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

405426-1G 405426-5G
SIAL405426-1GEA 51 EUR
SIAL405426-1G SIAL405426-5G
2,6-Bis(brommethyl)pyridin, Sigma-Aldrich®
2,6-Bis(brommethyl)pyridin
Formel: C₇H₇Br₂N
Molecular Weight: 264,95 g/mol
MDL: MFCD00191795
CAS-Nummer: 7703-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR