Benzyl-3-(Cbz-amino)picolinat

Lieferant: COMBI-BLOCKS

HC-3400-5G HC-3400-1G
COBBHC-3400-5GEA 595 EUR
COBBHC-3400-5G COBBHC-3400-1G
Benzyl-3-(Cbz-amino)picolinat
Benzyl-3-(Cbz-amino)picolinat
Formel: C₂₁H₁₈N₂O₄
MDL: MFCD16618953
CAS-Nummer: 1228956-95-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR