N-(4-Methoxybenzyl)-5-brompicolinamid
Lieferant: COMBI-BLOCKS
Synonyme:
5-Bromo-N-(4-methoxybenzyl)picolinamide, 5-Bromo-N-(4-methoxybenzyl)pyridine-2-carboxamide
PY-7208-1G
PY-7208-5G
COBBPY-7208-1GEA
315
EUR
COBBPY-7208-1G
COBBPY-7208-5G
N-(4-Methoxybenzyl)-5-brompicolinamid
N-(4-Methoxybenzyl)-5-brompicolinamid
Formel:
C₁₄H₁₃BrN₂O₂ Molecular Weight: 321,1692 g/mol |
MDL:
MFCD09801060 CAS-Nummer: 951885-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...