2-Amino-4-methylthiophen-3-carbonsäureamid
Lieferant: FLUOROCHEM
24813-25G
024813-25G
FLUO24813-25GEA
0
EUR
FLUO24813-25G
FLUO024813-25G
2-Amino-4-methylthiophen-3-carbonsäureamid
2-Amino-4-methylthiophene-3-carboxamide
Formel:
C₆H₈N₂OS Molecular Weight: 156,20 g/mol |
MDL:
MFCD00052594 CAS-Nummer: 4651-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...