4-(2-Aminoethyl)thiomorpholin

Lieferant: COMBI-BLOCKS

AM-1002-1G AM-1002-5G
COBBAM-1002-1GEA 0 EUR
COBBAM-1002-1G COBBAM-1002-5G
4-(2-Aminoethyl)thiomorpholin
4-(2-Aminoethyl)thiomorpholin
Formel: C₆H₁₄N₂S
MDL: MFCD02179434
CAS-Nummer: 53515-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR