5-Chlor-4-chinazolon
Lieferant: COMBI-BLOCKS
Synonyme:
5-Chloro-3H-quinazolin-4-one, 5-Chloro-3,4-dihydro-4-oxoquinazoline
HC-6061-1G
HC-6061-25G
HC-6061-5G
COBBHC-6061-1GEA
101
EUR
COBBHC-6061-1G
COBBHC-6061-25G
COBBHC-6061-5G
5-Chlor-4-chinazolon
5-Chlor-4-chinazolon
Formel:
C₈H₅ClN₂O Molecular Weight: 180,5911 g/mol |
MDL:
MFCD02676498 CAS-Nummer: 60233-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...