6-Chlor-4-chinazolon

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chloroquinazolin-4-ol

HC-6040-5G HC-6040-1G
COBBHC-6040-5GEA 181 EUR
COBBHC-6040-5G COBBHC-6040-1G
6-Chlor-4-chinazolon
6-Chlor-4-chinazolon
Formel: C₈H₅ClN₂O
Molecular Weight: 180,5900 g/mol
MDL: MFCD01686365
CAS-Nummer: 16064-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR