5-Fluor-4-chinazolon

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluoroquinazolin-4(1H)-one, 5-Fluoro-4-hydroxyquinazoline

HC-6057-1G HC-6057-5G
COBBHC-6057-1GEA 38.2 EUR
COBBHC-6057-1G COBBHC-6057-5G
5-Fluor-4-chinazolon
5-Fluor-4-chinazolon
Formel: C₈H₅FN₂O
Molecular Weight: 164.13 g/mol
MDL: MFCD09475836
CAS-Nummer: 436-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR