6-Nitro-4-chinazolon

Lieferant: COMBI-BLOCKS

Synonyme: 6-Nitroquinazolin-4(3H)-one, 6-Nitro-4(3H)-quinazolinone

HC-6569-1G HC-6569-5G
COBBHC-6569-1GEA 127 EUR
COBBHC-6569-1G COBBHC-6569-5G
6-Nitro-4-chinazolon
6-Nitro-4-chinazolon
Formel: C₈H₅N₃O₃
Molecular Weight: 191,1436 g/mol
MDL: MFCD00137845
CAS-Nummer: 6943-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR