2-Morpholinobenzaldehyd
Lieferant: Thermo Scientific
SEW05383DA
SEW05383EA
MAYBSEW05383DAEA
0
EUR
MAYBSEW05383DA
MAYBSEW05383EA
2-Morpholinobenzaldehyd
2-Morpholinobenzaldehyd
Formel:
C₁₁H₁₃NO₂ Molecular Weight: 191,2300 g/mol |
MDL:
MFCD00662562 CAS-Nummer: 58028-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...