5-(4-Morpholinyl)-2-nitroanilin
Lieferant: BIONET RESEARCH
3T-0352.500
3T-0352.1
3T-0352.5
BIOT3T-0352.500EA
0
EUR
BIOT3T-0352.500
BIOT3T-0352.1
BIOT3T-0352.5
5-(4-Morpholinyl)-2-nitroanilin
5-(4-Morpholinyl)-2-nitroanilin
Formel:
C₁₀H₁₃N₃O₃ |
MDL:
MFCD00511844 CAS-Nummer: 54998-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...