N-[(Benzyloxy)carbonyl]-D-tryptophan
Lieferant: BACHEM BIOCHEMICA
Synonyme:
Z-D-Trp-OH, N(α)-Benzyloxycarbonyl-D-tryptophan
4001610.0005
4001610.0025
BACE4001610.0005EA
0
EUR
BACE4001610.0005
BACE4001610.0025
N-[(Benzyloxy)carbonyl]-D-tryptophan
N(α)-Cbz-D-tryptophan
Formel:
C₁₉H₁₈N₂O₄ Molecular Weight: 338,4 g/mol |
MDL:
MFCD00037945 CAS-Nummer: 2279-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...