N-[(Benzyloxy)carbonyl]-L-tryptophan

Lieferant: BACHEM BIOCHEMICA

Synonyme: (S)-2-(((Benzyloxy)carbonyl)amino)-3-(1H-indol-3-yl)propanoic acid, N(α)-Benzyloxycarbonyl-L-tryptophan, N-Cbz-L-Tryptophan, Z-Trp-OH

4001218.0025 4001218.0100
BACE4001218.0025EA 0 EUR
BACE4001218.0025 BACE4001218.0100
N-[(Benzyloxy)carbonyl]-L-tryptophan
N(α)-Cbz-L-tryptophan
Formel: C₁₉H₁₈N₂O₄
Molecular Weight: 338.4 g/mol
MDL: MFCD00065700
CAS-Nummer: 7432-21-5
EINECS: 231-074-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR