(Tetrahydropyran-4-yliden)acetonitril
Lieferant: COMBI-BLOCKS
Synonyme:
2-(Dihydro-2H-pyran-4(3H)-ylidene)acetonitrile
AM-1621-5G
AM-1621-1G
COBBAM-1621-5GEA
0
EUR
COBBAM-1621-5G
COBBAM-1621-1G
(Tetrahydropyran-4-yliden)acetonitril
(Tetrahydropyran-4-yliden)acetonitril
Formel:
C₇H₉NO |
MDL:
MFCD09800849 CAS-Nummer: 204651-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...