2-(1-Piperidinyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
PN-5476-5G
PN-5476-1G
COBBPN-5476-5GEA
0
EUR
COBBPN-5476-5G
COBBPN-5476-1G
2-(1-Piperidinyl)phenylboronsäurepinakolester
2-(1-Piperidinyl)phenylboronsäurepinakolester
Formel:
C₁₇H₂₆BNO₂ |
MDL:
MFCD07368528 CAS-Nummer: 852227-97-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...