4-(Piperidin-1-carbonyloxy)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl piperidine-1-carboxylate
PN-3597-1G
PN-3597-5G
COBBPN-3597-1GEA
0
EUR
COBBPN-3597-1G
COBBPN-3597-5G
4-(Piperidin-1-carbonyloxy)phenylboronsäurepinakolester
4-(Piperidin-1-carbonyloxy)phenylboronsäurepinakolester
Formel:
C₁₈H₂₆BNO₄ |
MDL:
MFCD09027287 CAS-Nummer: 913836-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...