4-(2-(Piperidin-1-yl)ethoxy)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethyl)piperidine

PN-3800-5G PN-3800-1G
COBBPN-3800-5GEA 1850 EUR
COBBPN-3800-5G COBBPN-3800-1G
4-(2-(Piperidin-1-yl)ethoxy)phenylboronsäurepinakolester
4-(2-(Piperidin-1-yl)ethoxy)phenylboronsäurepinakolester
Formel: C₁₉H₃₀BNO₃
Molecular Weight: 331,26 g/mol
MDL: MFCD09258652
CAS-Nummer: 934586-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR