3-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-((2-(Piperidin-1-yl)ethyl)carbamoyl)phenyl)boronic acid

BB-5260-5G BB-5260-1G
COBBBB-5260-5GEA 1960 EUR
COBBBB-5260-5G COBBBB-5260-1G
3-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronsäure
3-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronsäure
Formel: C₁₄H₂₁BN₂O₃
Molecular Weight: 276,1391 g/mol
MDL: MFCD11053842
CAS-Nummer: 1072946-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR