4-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronsäure Hydrochlorid
Lieferant: COMBI-BLOCKS
BB-3218-1G
BB-3218-5G
COBBBB-3218-1GEA
63.9
EUR
COBBBB-3218-1G
COBBBB-3218-5G
4-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronsäure Hydrochlorid
4-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronsäure Hydrochlorid
Formel:
C₁₄H₂₂BClN₂O₃ |
MDL:
MFCD09027221 CAS-Nummer: 957060-72-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...