3-Nitro-4-(1-piperidinyl)benzaldehyd

Lieferant: BIONET RESEARCH

4Y-0715.500 4Y-0715.5 4Y-0715.1 4Y-0715.10
BIOT4Y-0715.500EA 0 EUR
BIOT4Y-0715.500 BIOT4Y-0715.5 BIOT4Y-0715.1 BIOT4Y-0715.10
3-Nitro-4-(1-piperidinyl)benzaldehyd
3-Nitro-4-(1-piperidinyl)benzaldehyd
Formel: C₁₂H₁₄N₂O₃
Molecular Weight: 234.26 g/mol
MDL: MFCD00831780
CAS-Nummer: 39911-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR