Ethyl-1-(4-bromphenyl)cyclopropancarboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 1-(4-bromophenyl)cyclopropanecarboxylate

HC-3049-5G HC-3049-1G
COBBHC-3049-5GEA 608 EUR
COBBHC-3049-5G COBBHC-3049-1G
Ethyl-1-(4-bromphenyl)cyclopropancarboxylat
Ethyl-1-(4-bromphenyl)cyclopropancarboxylat
Formel: C₁₂H₁₃BrO₂
Molecular Weight: 269.1300 g/mol
MDL: MFCD15143548
CAS-Nummer: 1215205-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR