Methyl-1-(4-bromphenyl)cyclopropancarboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 1-(4-bromophenyl)cyclopropanecarboxylate

HC-3048-5G HC-3048-1G
COBBHC-3048-5GEA 820 EUR
COBBHC-3048-5G COBBHC-3048-1G
Methyl-1-(4-bromphenyl)cyclopropancarboxylat
Methyl-1-(4-bromphenyl)cyclopropancarboxylat
Formel: C₁₁H₁₁BrO₂
Molecular Weight: 255.11 g/mol
MDL: MFCD12913964
CAS-Nummer: 638220-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR