1-(3-Bromphenyl)cyclopropancarbonsäure

Lieferant: COMBI-BLOCKS

HC-2890-5G HC-2890-1G
COBBHC-2890-5GEA 282 EUR
COBBHC-2890-5G COBBHC-2890-1G
1-(3-Bromphenyl)cyclopropancarbonsäure
1-(3-Bromophenyl)cyclopropanecarboxylic acid
Formel: C₁₀H₉BrO₂
Molecular Weight: 241,0813 g/mol
MDL: MFCD07374441
CAS-Nummer: 124276-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR