1-(4-Chlorphenyl)cyclopropancarbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid

HC-2916-25G HC-2916-5G HC-2916-1G
COBBHC-2916-25GEA 288 EUR
COBBHC-2916-25G COBBHC-2916-5G COBBHC-2916-1G
1-(4-Chlorphenyl)cyclopropancarbonsäure
1-(4-Chlorophenyl)cyclopropanecarboxylic acid
Formel: C₁₀H₉ClO₂
Molecular Weight: 196,64 g/mol
MDL: MFCD00001289
CAS-Nummer: 72934-37-3
EINECS: 277-105-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR