1-(4-Methoxyphenyl)cyclopropancarbonsäure

Lieferant: COMBI-BLOCKS

HC-2999-5G HC-2999-25G HC-2999-1G
COBBHC-2999-5GEA 0 EUR
COBBHC-2999-5G COBBHC-2999-25G COBBHC-2999-1G
1-(4-Methoxyphenyl)cyclopropancarbonsäure
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid
Formel: C₁₁H₁₂O₃
MDL: MFCD00019223
CAS-Nummer: 16728-01-1
EINECS: 240-788-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR